LABORATORY FREEZE-DRYING UNIT VERSION 11+Manifold with valves/ 120V 60Hz Model 1 LYO GT 2-B Basic Unit This unit is provided with a pressure indicator over handheld display, compressor, stainless steel condenser, connecting cable vacuum hose. Vacuum 1 * 1 items
Catalog # SRKALYOGT2-B
Supplier: SRK SYSTEMTECHNIK GMBH
Frequently Bought Together