Order Entry
France
ContactUsLinkComponent
Neutral field screen set to correct image background contains 1 x blue screen and 1 x frosted screen 335mm x 330mm x 15mm for use with G:BOX systems 1 * 1 items
Catalog # SYNOGX-NFS
Neutral field screen set to correct image background contains 1 x blue screen and 1 x frosted screen 335mm x 330mm x 15mm for use with G:BOX systems 1 * 1 items
Catalog # SYNOGX-NFS
Frequently Bought Together