Water for ionic chromatography 1 * 1.000 mL
Catalog # ACSDCIW.1L
Supplier: CPACHEM
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR® Conductivity standard, CRM, 5000 μS/cm
Supplier: VWR Chemicals
Benzoic acid, Supelco®
Supplier: Merck
VWR®, Osmolality standards
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
BUFFER PH 10 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ