Gas40h2s,100 co,2.5% ch4,15% o2,n2 58l 4ga15x25w40h100-58 1 * 1 items
BWTE4GA15X25W40H10
Gas40h2s,100 co,2.5% ch4,15% o2,n2 58l 4ga15x25w40h100-58 1 * 1 items
BWTE4GA15X25W40H10
Frequently Bought Together
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection