Order Entry
France
ContactUsLinkComponent
Elite-bac advantage columns are exceptionally robust, ensuring longer column lifetime and highly reproducible data for analysis of b lood alcohol content (bac). inner diameter (mm i.d): 0.53 length (m): 30 film thickness (?m): 1.00. key benefits incl 1 * 1 items
Catalog # PERCN9315074
Supplier:  PerkinElmer
Elite-bac advantage columns are exceptionally robust, ensuring longer column lifetime and highly reproducible data for analysis of b lood alcohol content (bac). inner diameter (mm i.d): 0.53 length (m): 30 film thickness (?m): 1.00. key benefits incl 1 * 1 items
Catalog # PERCN9315074
Supplier:  PerkinElmer
Frequently Bought Together