Order Entry
France
ContactUsLinkComponent
kit, insulin (sheep) elisa kit, insulin (sheep) elisa kit is an immunoassay for the quantitative determination of circulating insuli n, a peptide hormone in sheep serum, plasma. 1 * 1 KIT
Catalog # ABNOKA2321
Supplier:  Abnova
kit, insulin (sheep) elisa kit, insulin (sheep) elisa kit is an immunoassay for the quantitative determination of circulating insuli n, a peptide hormone in sheep serum, plasma. 1 * 1 KIT
Catalog # ABNOKA2321
Supplier:  Abnova
Frequently Bought Together