Order Entry
France
ContactUsLinkComponent
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.7339g/ml @ 40C 1 * 100 ml
Catalog # REGEDEN40220
Supplier:  REAGECON
Free from toxic heavy metals in formulation. Manufactured to meet ASTM protocols. Tested, certified & batch traceable Density Standa rd 1.7339g/ml @ 40C 1 * 100 ml
Catalog # REGEDEN40220
Supplier:  REAGECON
Frequently Bought Together