Order Entry
France
Orders LinkContactUsLinkComponent
 

Cut resistant gloves, Sharpflex Latex

10, Yellow/blue, 250 mm
Catalog # DALL2232525/10
Cut resistant gloves, Sharpflex Latex
Catalog # DALL2232525/10
Cut resistant gloves, Sharpflex Latex
Cut resistant gloves, Sharpflex Latex

10, Yellow/blue, 250 mm

Price
15,19 €
Log in for availability
1 * 1 PAIRE
Product Details & Documents

These gloves knitted from composite and para-aramid yarn features a blue crinkled latex coating on the palm.

  • Knitting technique provides the glove with excellent fit and high dexterity
  • Para-aramid yarn protects against contact heat and cuts
  • Plated polyamide inside: Limit the contact of the skin with composite fibre
  • Latex coating provides better grip and light thermal protection (up to 250 °C)

Ideal for handling heavy parts with high cut risks in humid and slippery environments. The gloves also provide light thermal protection (up to 250 °C).

Certifications: EN 388:2016 (2X42C), EN 407 (X2XXXX); Cat. II

Specifications
  • Glove size:
    10
  • Colour:
    Yellow/blue
  • Length:
    250 mm
  • Seq:
    04
  • Cat. No.:
    DALL2232525/10
  • Liner material:
    Para-aramid
  • Disposable / reusable:
    Reusable
  • Model:
    Latex
  • Cuff style:
    Knit wrist
  • EN norm:
    EN 388:2016 (2X42C), EN 407 (X2XXXX)
  • Finish:
    Crinkled coating on palm and fingertips
  • Ambidextrous / hand-specific:
    Hand-specific
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.