Single channel pipettes, electronic, variable volume, E4™ XLS™
500 - 5000 µl, 5,0 µl, 2,4 - 0,6 %, 0,6 - 0,16 %, LTS™
RAIN17012312
Marketsource
: RAININ
The E4™ XLS™ is an electronic pipette that can be upgraded and expanded right from the PC. Available as standard versions or with LTS LiteTouch System for lower ejection forces.
- Large screen, joystick control and carousel menus speed access to all modes and options
- True Manual™ pipetting mode permits continuous Real Time control of pipetting speed and direction, combined with precise volume measurement
- Independent control of aspirate, dispense and mix speeds for challenging liquids
- Blowout and automatic dispensing options for precision pipetting protocols
- Ergonomic fingerhook and joystick provide maximum comfort
Standard functions include basic pipetting, multi-dispensing, and mixing. Advanced functions include sequential volume programming, fixed volume, titration, dilution and reverse mode pipetting.
: Supplied with Li-Ion battery and wall power supply. An optional charging stand is available separately.
- Volume:500 - 5000 µl
- With:LTS™
- Seq:08
- Increment:5,0 µl
- Accuracy:2,4 - 0,6 %
- Imprecision:0,6 - 0,16 %



