Order Entry
France
Orders LinkContactUsLinkComponent
Safety helmets, Concept HeightMaster

Safety helmets, Concept HeightMaster

Safety helmet, vented, without ratchet, EU, Black
Catalog # CENUS08EKFM
Safety helmets, Concept HeightMaster
Catalog # CENUS08EKFM
Safety helmets, Concept HeightMaster
Safety helmets, Concept HeightMaster

Safety helmet, vented, without ratchet, EU, Black

Price
Demander un prix
Connexion pour la disponibilité
1 * 1 Pce
Product Details & Documents

Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene). The Concept HeightMaster has been designed for applications where the wearer is working at height, where the suspension system and the helmet design and structure will reduce the risk of the helmet coming off as a result of an impact or a fall.

  • With extended nape area
  • Six point terylene inner cradle
  • Highly absorbent hydro-flock sweatband
  • With increased side rigidity (additional lateral deformity test)
  • Ventilation through wide ventilation slots
  • With or without ratchet, with full peak

Size: 510 to 630 mm

Weight: 325 g

Certifications: EN 397, EN 12492, EN 50365

Spécifications
  • Colour:
    Black
  • Description:
    Safety helmet, vented, without ratchet, EU
  • Seq:
    04
Accessories

Informations sur le produit

  • Catalog #
    Availability
    Price
  • VentPlugs, for closing ventilation holes, for Concept (vented) helmet
    VentPlugs, for closing ventilation holes, for Concept (vented) helmet
    CENUS30VP
    Connexion pour la disponibilité
    1 * 1 Pce

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Sweatband S31P, PVC, for Concept helmet
    Sweatband S31P, PVC, for Concept helmet
    CENUS31P
    Connexion pour la disponibilité
    1 * 1 Pce - 2,30 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.