8 COMPOSES 1000 MG/L CHAQUE Fluorides (F-) Chlorides (Cl-) Nitrites (NO2-) Nitrates (NO3-) Bromides (Br-) Phosphates (PO43-) Sulphates (SO42-) Iodides (I-) DANS H2O 1 * 100 mL
Catalog # 89253.180
Supplier: VWR Collection
Frequently Bought Together