Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Glass bottle, 250 mL
Supplier: Avantor
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 8025.0250
Supplier: Avantor
CAS Number: 64-17-5
Jump to:
Product Details & Documents
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Merck Index:12,03806
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Documents
Specifications
Specification Test Results
- Assay:min. 99,9 % (v/v)
- Acetone, isopropylalcohol:passes test
- Color (APHA):max. 10
- Methanol (CH3OH):max. 0,1 %
- Residue after Evaporation:max. 0,001 %
- Solubility in Water:passes test
- Substances Darkened by H2SO4:passes test
- Substances Reducing KMnO4:passes test
- Titrable Acid (meq/g):max. 0,0005
- Titrable Base (meq/g):max. 0,0002
- Water (H2O):max. 0,2 %
- Trace Impurities (in ppm):-
- Aluminium (Al):max. 0,5
- Barium (Ba):max. 0,1
- Boron (B):max. 0,02
- Cadmium (Cd):max. 0,05
- Calcium (Ca):max. 0,5
- Chromium (Cr):max. 0,02
- Cobalt (Co):max. 0,02
- Copper (Cu):max. 0,2
- Iron (Fe):max. 0,1
- Lead (Pb):max. 0,1
- Magnesium (Mg):max. 0,1
- Manganese (Mn):max. 0,02
- Nickel (Ni):max. 0,02
- Tin (Sn):max. 0,1
- Zinc (Zn):max. 0,1
Specifications
- Pack type:Glass bottle
- Pk:250 mL
- Cat. No.:8025.0250
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



