Saisie de commande
France
Lien pour commanderContactUsLinkComponent
Sodium Picosulfate, Reference Standards

Sodium Picosulfate, Reference Standards

Sodium Picosulfate Related Compound A, USP reference standard, 20 mg
Référence 1614589.
Fournisseur:   USP
Sodium Picosulfate, Reference Standards
Référence 1614589.
Fournisseur:  USP
Numéro CAS:  
Sodium Picosulfate, Reference Standards

Sodium Picosulfate Related Compound A, USP reference standard, 20 mg

Numéro CAS:  
Prix
1 190,00 €
Log in for availability
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Cdt:
    20 mg
  • Normes:
    USP reference standard
  • Désignation:
    Sodium Picosulfate Related Compound A
  • Cat. No.:
    1614589.
  • N ° CAS.:
    32500-19-9
  • Désignation:
    Sodium Picosulfate Related Compound A
  • Synonyme(s):
    Sodium 4-((4-hydroxyphenyl)(pyridin-2-yl)methyl)phenyl sulfate

Specifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.