Order Entry
France
Orders LinkContactUsLinkComponent
X-Type syringes for sensitive chromatography applications

X-Type syringes for sensitive chromatography applications

22s, 100 µl, 51 mm, 0,72 mm, 3, 1710 FN CTC, X-Type
Catalog # 549-0696
Supplier:   HAMILTON BONADUZ
X-Type syringes for sensitive chromatography applications
Catalog # 549-0696
Supplier:  HAMILTON BONADUZ
X-Type syringes for sensitive chromatography applications
X-Type syringes for sensitive chromatography applications

22s, 100 µl, 51 mm, 0,72 mm, 3, 1710 FN CTC, X-Type

Price
260,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents

X-Type syringes are perfectly suited for sensitive LC samples such as proteins, peptides, phospholipids or smaller amines.

The syringe features a deactivated glass barrel and needle with an inorganic coating to prevent adsorption on the inner surfaces, resulting in nearly zero sample carryover. The coating creates a smooth inner barrel surface, leading to increased syringe lifetime by lowering friction during high speed applications while maintaining a perfect seal over the entire life of the syringe. The deactivated X-Type needle is directly attached to the barrel without using cement, further reducing the risk of sample carryover.

Fixed needle (FN) - This termination is found on CTC chromatography syringes. The unique design features a direct attachment of the needle to the barrel and eliminates contact between the sample solvent and the adhesive reducing carryover.

X-Type syringes are specially designed for the CTC LC PAL, HTC PAL, HTS PAL HTX PAL, PAL HTC-xt, PAL HTS-xt and PAL HTX-xt extended autosamplers sold under the following brands like AB Sciex, Agilent, Alpha M.O.S., Antek, Bruker, Dionex, GE, Gerstel, GL Sciences, Lauda, LEAP Technologies, MicroCal, MPS, Perichrom, Perkin Elmer, Shimadzu, Sotax, Thermo Scientific, Waters and Zoex.

Specifications
  • Ext.Ø:
    0,72 mm
  • Gauge:
    22s
  • Type:
    1710 FN CTC, X-Type
  • Type of tip:
    3
  • Volume:
    100 µl
  • Length:
    51 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.