Sampling spatula, Sterileware®, SP Bel-Art
+, 229 mm, 153 mm, 15 ml, White
Supplier: Bel-Art Products, a Part of SP
Sampling spatula, Sterileware®, SP Bel-Art
Catalog # 231-0014
Supplier: Bel-Art Products, a Part of SP
Jump to:
Product Details & Documents
One-piece spatulas, moulded from FDA grade, high impact PS.
- Sterilised by gamma irradiation, lot stamped for accurate lot tracing, shipped with a Certificate of Processing for Sterilisation
V-shaped spatula for sampling dislodge caked or crystallised reagents from deep within tall bottles or vessels having a neck diameter as small as 18 mm (³/₄"). The sturdy point is perfect for taking sterile samples during process quality inspections, and for piercing plastic-lined paper bags or similar packages.
Documents
Specifications
- Sterile:+
- Capacity:15 ml
- Colour:White
- Length:229 mm
- Seq:01
- Blade length:153 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



