Order Entry
France
Orders LinkContactUsLinkComponent
Multi-channel pipettes, electronic, variable volume, E4™ XLS™
Multi-channel pipettes, electronic, variable volume, E4™ XLS™
100 - 1200 µl, 1,0 µl, ±3,6 (for 100 µl); ±0,8 (for 600 µl), ±0,8 (for 1200 µl) %, ≤0,6 (100 µl); ≤0,2 (600 µl), ≤0,15 (1200 µl) %, RFID tag
Catalog # RAINE8-1200XLS
Marketsource
Supplier:   RAININ
Multi-channel pipettes, electronic, variable volume, E4™ XLS™
Catalog # RAINE8-1200XLS
Supplier:  RAININ
Marketsource
Multi-channel pipettes, electronic, variable volume, E4™ XLS™
100 - 1200 µl, 1,0 µl, ±3,6 (for 100 µl); ±0,8 (for 600 µl), ±0,8 (for 1200 µl) %, ≤0,6 (100 µl); ≤0,2 (600 µl), ≤0,15 (1200 µl) %, RFID tag
Price
2 910,00 €
Log in for availability
1 * 1 Un.
Product Details & Documents
Fast loading, easy to handle multi-channel pipette for high-throughput work.
  • Menu structure, joystick control and large colour screen make switching between modes and setting functions quick and easy
  • LTS’s cylindrical tip design assures fast tip mounting, consistent sample loading and leak-free seals across all channels
  • Autopace simplifies repeat dispensing – click the joystick once and repeat dispense across a plate
  • Independent adjustments for aspirate, dispense and mix speeds accommodate challenging liquids
  • Blowout and automatic dispensing options enable precision pipetting protocols
  • Ergonomically designed fingerhook and joystick for maximum comfort
  • Easily accessible service records and alarms
  • RFID enabled for advanced asset management
Standard functions include Basic pipetting, Multi-dispense, Mixing and True Manual™ pipetting mode. Advanced functions include sequential volume programming, fixed volume pipetting, titration, dilution and reverse mode pipetting.
Ordering information: Supplied with Li-Ion battery and wall power supply. Rapid charge stand is optionally available.
Specifications
  • Volume:
    100 - 1200 µl
  • With:
    RFID tag
  • Seq:
    01
  • Increment:
    1,0 µl
  • Accuracy:
    ±3,6 (for 100 µl); ±0,8 (for 600 µl), ±0,8 (for 1200 µl) %
  • Imprecision:
    ≤0,6 (100 µl); ≤0,2 (600 µl), ≤0,15 (1200 µl) %