Multi-channel pipettes, electronic, variable volume, E4™ XLS™
100 - 1200 µl, 1,0 µl, ±3,6 (for 100 µl); ±0,8 (for 600 µl), ±0,8 (for 1200 µl) %, ≤0,6 (100 µl); ≤0,2 (600 µl), ≤0,15 (1200 µl) %, RFID tag
Catalog # RAINE12-1200XLS
Marketsource
Supplier: RAININ
Multi-channel pipettes, electronic, variable volume, E4™ XLS™
Catalog # RAINE12-1200XLS
Supplier: RAININ
Marketsource
Jump to:
Product Details & Documents
Fast loading, easy to handle multi-channel pipette for high-throughput work.
- Menu structure, joystick control and large colour screen make switching between modes and setting functions quick and easy
- LTS’s cylindrical tip design assures fast tip mounting, consistent sample loading and leak-free seals across all channels
- Autopace simplifies repeat dispensing – click the joystick once and repeat dispense across a plate
- Independent adjustments for aspirate, dispense and mix speeds accommodate challenging liquids
- Blowout and automatic dispensing options enable precision pipetting protocols
- Ergonomically designed fingerhook and joystick for maximum comfort
- Easily accessible service records and alarms
- RFID enabled for advanced asset management
Standard functions include Basic pipetting, Multi-dispense, Mixing and True Manual™ pipetting mode. Advanced functions include sequential volume programming, fixed volume pipetting, titration, dilution and reverse mode pipetting.
Ordering information: Supplied with Li-Ion battery and wall power supply. Rapid charge stand is optionally available.
Specifications
- Volume:100 - 1200 µl
- With:RFID tag
- Seq:03
- Increment:1,0 µl
- Accuracy:±3,6 (for 100 µl); ±0,8 (for 600 µl), ±0,8 (for 1200 µl) %
- Imprecision:≤0,6 (100 µl); ≤0,2 (600 µl), ≤0,15 (1200 µl) %