Order Entry
France
ContactUsLinkComponent
RECOMBINANT/NATIVE PROTEIN, CSF1 (HUMAN) RECOMBINANT PROTEIN, HUMAN CSF1 (P09603, 33 A.A. - 181 A.A.) PARTIAL RECOMBINANT PROTEIN EX PRESSED IN BACULOVIRUS INFECTED BOMBYX MORI. 1 * 1 Each
Catalog # ABNOP3649
Supplier:  Abnova
RECOMBINANT/NATIVE PROTEIN, CSF1 (HUMAN) RECOMBINANT PROTEIN, HUMAN CSF1 (P09603, 33 A.A. - 181 A.A.) PARTIAL RECOMBINANT PROTEIN EX PRESSED IN BACULOVIRUS INFECTED BOMBYX MORI. 1 * 1 Each
Catalog # ABNOP3649
Supplier:  Abnova