Order Entry
France
ContactUsLinkComponent
Calibration Slide Universal D=0 without coverslip positive version - high-precision object relocation - position of reference mark relative to left and upper slide border X=38mm/Y=13mm, +/- 5µm - line with 50mm length to control parallelism - Stage 1 * 1 items
Catalog # ZEIS4740299010000
Supplier:  ZEISS
Calibration Slide Universal D=0 without coverslip positive version - high-precision object relocation - position of reference mark relative to left and upper slide border X=38mm/Y=13mm, +/- 5µm - line with 50mm length to control parallelism - Stage 1 * 1 items
Catalog # ZEIS4740299010000
Supplier:  ZEISS
Frequently Bought Together