Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4
TBS buffer solution 10X concentrate pH 7,4, 500 mL
Catalog # CAYM600232-500
Supplier: Cayman Chemical
Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4
Catalog # CAYM600232-500
Supplier: Cayman Chemical
CAS Number:
Jump to:
Product Details & Documents
Formulation: Concentrated TBS (250 mM Tris-base; pH 7,4; 1,37 M sodium chloride and 27 mM KCl).
Suggested storage (long term) temperature: 4 °C.
Documents
Specifications
Specifications
- Pk:500 mL
- Description:TBS buffer solution 10X concentrate pH 7,4



