Order Entry
France
ContactUsLinkComponent
LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 25MM/330-450MM, LRC25X330-450V01 1 * 1 items
Catalog # 554-2382
undefined
LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 25MM/330-450MM, LRC25X330-450V01 1 * 1 items
Catalog # 554-2382