Order Entry
France
ContactUsLinkComponent
LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 10MM/0-120MM, LRC10X000-120V01 1 * 1 items
Catalog # 554-2363
undefined
LRC columns offer 4 internal diameters and various lengths to provide a range of bed heights and column volumes. Made of borosilicat e glass and equipped with one adjustable and one fixed piston. LRC COLUMN 10MM/0-120MM, LRC10X000-120V01 1 * 1 items
Catalog # 554-2363