Order Entry
France
Orders LinkContactUsLinkComponent
Hydrochloric acid 5%

Hydrochloric acid 5%

Glass bottle, 1 L
Catalog # PRLS88942.290
Corrosive
Supplier:   PROLABO CMP
Hydrochloric acid 5%
Catalog # PRLS88942.290
Supplier:  PROLABO CMP
CAS Number:  7647-01-0
Hydrochloric acid 5%

Glass bottle, 1 L

CAS Number:  7647-01-0
Price
42,20 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:∼100 °C (1013 hPa)
  • Density:1.02…1.05 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.