Tablets Phosphate Silica Reducing 1 * 250 items
Catalog # PALIAT116
Supplier: Palintest
Frequently Bought Together
BE 20MG/L IN HNO3 2% HF 0.5% 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
STREPTOMYCIN ELISA KIT 1 * 1 KIT
Supplier: ENZO LIFE SCIENCES
SODIUIM HYDROXIDE 30% SOLUTION 1 * 500 mL
Supplier: Spectrum Chemical
Reagents for Phosphate Determination using Lovibond® MD photometers
Supplier: Lovibond
trans-4-Hydroxycinnamic acid ≥98% (by HPLC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
HUMAN HAIR FOLLICLE STEM CELL COMPLETE MEDIA WITH SERUM 1 * 500 mL
Supplier: MARKET SOURCE PART PROCESS
KJELTABS KPC 1 * 1.000 Tablet
Supplier: THOMPSON & CAPPER
Sodium hydroxide, anhydrous ≥98%, pellets, Reagent Grade
Supplier: Honeywell Chemicals
Test kits, phosphate (kits and reagents) CHECKIT®
Supplier: Lovibond
VWR®, Powder packs for determination of chlorine, reagents for photometers
Supplier: VWR Chemicals