0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Sulphide Test 0-0.5Mgl Pk50 Calibration Chart Palintest 1 * 50 items
PALIPM168
:
Palintest
Sulphide Test 0-0.5Mgl Pk50 Calibration Chart Palintest 1 * 50 items
PALIPM168
:
Palintest
Frequently Bought Together
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test kits, photometric, ammonium, LCK cuvette tests
: Hach
Test strips, sulphite, MQuant®, Supelco®
: Merck
Schiff reagent for histology, Sigma-Aldrich®
: Merck
Test strips, nitrate/nitrite, Dosatest
: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
: Merck
Test strips for free chlorine, MQuant®, Supelco®
: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Sodium hydroxide 10% in aqueous solution, EMSURE® for analysis, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
May Grunwald's eosin-methylene blue modified solution stain for hematology, for cytology, Sigma-Aldrich®
: Merck