Order Entry
France
ContactUsLinkComponent
inoLab pH 7110 BNC SET 7:Easy-to-operate basic pH/mV benchtop meter with BNC socket for routine measurement. For AC and battery ope ration. Meter with universal power supply, stand and operation manual. Combined pH electrode SenTix 42, buffer 4, 7 1 * 1 items
Catalog # 662-1360
Supplier:  WTW
inoLab pH 7110 BNC SET 7:Easy-to-operate basic pH/mV benchtop meter with BNC socket for routine measurement. For AC and battery ope ration. Meter with universal power supply, stand and operation manual. Combined pH electrode SenTix 42, buffer 4, 7 1 * 1 items
Catalog # 662-1360
Supplier:  WTW
Frequently Bought Together