Order Entry
France
Orders LinkContactUsLinkComponent
Ventilation unit for storage cabinets

Ventilation unit for storage cabinets

Active coal filter for corrosive and organic vapours
Catalog # TRIOCDFCORG
Supplier:   TRIONYX
Ventilation unit for storage cabinets
Catalog # TRIOCDFCORG
Supplier:  TRIONYX
Ventilation unit for storage cabinets
Ventilation unit for storage cabinets

Active coal filter for corrosive and organic vapours

Price
210,12 €
Log in for availability
1 * 1 Pce
Product Details & Documents

Ventilation unit with control switch to adjust the fan air flow.

  • Airflow: 215 m³/hour
  • Dimension: 225×305×195 mm

Delivery information: Includes two outlets to attach to the ventilation of the cabinet and the unit and a flexible pipe to connect the unit to the cabinet.

Specifications
  • Description:
    Active coal filter for corrosive and organic vapours
  • Seq:
    004