Spatulas
300 mm
Catalog # BURK5313-0300
Supplier: BURKLE
Jump to:
Product Details & Documents
Stainless steel blade, wooden handle. Universal instrument for collecting, dividing and mixing pastes, creams etc.
Specifications
- Length:300 mm
- Seq:02