Precision thermometer, stem form ASTM 6 C, IP 2 C Low Cloud and Pour Capillaries form: yellow enamelled round liquid filling: Toluene red, range: -80+20°C in 1°C, immersion length: 76 mm, total length: 225 mm, suitable for calibration 1 * 1 items
Catalog # SCNL1202006
Supplier: Ludwig Schneider GmbH & CO. KG
Frequently Bought Together