Order Entry
France
ContactUsLinkComponent
DAD Package 2 (891-0112 Pump + 891-0212 AS w/ cooling + 891-0452 DAD detector + 891-0852 Organiser+890-0709 USB interface + HPLCLCUKIT2+ Damper case kit 891-1659) 1 * 1 SET
Catalog # HITA890-0613
Supplier:  VWR Collection
DAD Package 2 (891-0112 Pump + 891-0212 AS w/ cooling + 891-0452 DAD detector + 891-0852 Organiser+890-0709 USB interface + HPLCLCUKIT2+ Damper case kit 891-1659) 1 * 1 SET
Catalog # HITA890-0613
Supplier:  VWR Collection
Frequently Bought Together