Crystal violet solution
1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Primary stain for Gram staining
Synonyms: Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
- CAS Number:548-62-9
- Formula:C₂₅H₃₀ClN₃
- MDL Number:MFCD00011750
- Molecular Weight:407.99 g/mol
Documents
Certificates
Search by Lot Number911517ZA_VIOLET CRYSTAL SOLUTION_ISO 11133_rev1.pdf 911517ZAMP_VIOLET CRYSTAL SOLUTION_ISO 11133_rev1.pdf
spec_VWRC911517ZA
Product specification
VWR Microbiology
Product Literature
Specifications
Specifications
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



