Order Entry
France
Orders LinkContactUsLinkComponent
Nitromethane for synthesis, Sigma-Aldrich®

Nitromethane for synthesis, Sigma-Aldrich®

200 kg
Catalog # 8.20894.9200
Flammable
Health
Irritant
Supplier:   Merck
Nitromethane for synthesis, Sigma-Aldrich®
Catalog # 8.20894.9200
Supplier:  Merck
CAS Number:  75-52-5
Nitromethane for synthesis, Sigma-Aldrich®

200 kg

CAS Number:  75-52-5
Price
Inquire For Price
Log in for availability
1 * 200 kg
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:75-52-5
  • Formula:CH₃NO₂
  • MDL Number:MFCD00007400
  • Molecular Weight:61.04 g/mol
  • Merck Index:13,06644
  • Boiling Pt:101.2 °C (1013 hPa)
  • Melting Pt:–29 °C
  • Density:1.138 g/cm³ (20 °C)
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • UN:1261
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:200-876-6

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Solubility:
    105 g/l (20 °C)
  • Melting Point:
    -29 °C
  • Molar Mass:
    61.04 g/mol
  • Boiling Point:
    101.2 °C (1013 hPa)
  • Vapor Pressure:
    36.4 hPa (20 °C)
  • Flash Point:
    35.6 °C
  • Refractive Index:
    1.38056 (22 °C, 589 nm)
  • Explosion Limit:
    7.3 - 63.0 %(V)
  • Density:
    1.14 g/cm3 (20 °C)
  • pH:
    6.4 (0.6 g/l, H2O, 20 °C)
  • Ignition Point:
    418 °C
  • Saturation concentration:
    90 g/m3 (20 °C) Air
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specifications

  • Pk:
    200 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.