Order Entry
France
Orders LinkContactUsLinkComponent
Drying trays
Drying trays
400×300×40 mm
Catalog # JUCHVLSOBAYB807841
Drying trays
Catalog # JUCHVLSOBAYB807841
Drying trays
Drying trays
400×300×40 mm
Price
230,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Made of acid resistant stainless steel.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Specifications
  • Seq:
    03
  • W×D×H:
    400×300×40 mm