Order Entry
France
Orders LinkContactUsLinkComponent
Drying trays
Drying trays
180×130×40 mm
Catalog # JUCHVLSOBAYB807801
Drying trays
Catalog # JUCHVLSOBAYB807801
Drying trays
Drying trays
180×130×40 mm
Price
182,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents
Made of acid resistant stainless steel.
  • Very good chemical resistance
  • Very good thermal resistance
  • Smooth surfaces for easy cleaning
Specifications
  • Seq:
    01
  • W×D×H:
    180×130×40 mm