(±)-2-Hexanol ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 626-93-7 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 45°(113°F) Refractive Index: 1.4140 Odour: Characteristic Melting Point: -23° Boiling Point: 137-139° Beilstein Reference: 1718996 UN No.: 2282 MDL No.: MFCD00004585
Synonyms: (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
- CAS Number:626-93-7
- Formula:C₆H₁₄O
- MDL Number:MFCD00004585
- Molecular Weight:102.18 g/mol
- Boiling Pt:137…139 °C
- Melting Pt:–23 °C
- Density:0.810
- Flash Pt:45 °C (113 °F)
- Storage Temperature:Ambient
- EINECS:210-971-4
- UN:2282
- ADR:3,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



