Kit optical Fluorescence Glomax Multi Jr GFPUV (Ex 365nm, Em 515-570nm) 1 * 1 KIT
PROME6075
: Promega
Kit optical Fluorescence Glomax Multi Jr GFPUV (Ex 365nm, Em 515-570nm) 1 * 1 KIT
PROME6075
: Promega
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection