Chromatography, Acrodisc Units with Mustang Membranes Kit, Kit includes: 4 x Acrodisc units with Mustang Q membrane (non sterile) 4 x Acrodisc Unit with Mustang S membrane (non sterile) and 2 x Acrodisc unit with Mustang E membranes. 1 * 10 items
Catalog # 569-0020
Supplier: Cytiva (Formerly Pall Lab)
Frequently Bought Together