Order Entry
France
Orders LinkContactUsLinkComponent
HPLC columns

HPLC columns

Analytical, 2,10 mm, 150 mm
  JOURJR-65005NF
HPLC columns
  JOURJR-65005NF
HPLC columns

Analytical, 2,10 mm, 150 mm

 
 
   
1 * 1 Pce
 

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
 
  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Length:
    150 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.