Order Entry
France
Orders LinkContactUsLinkComponent
HPLC columns

HPLC columns

Analytical, 2,10 mm, 50 mm
  JOURJR-65002NF
HPLC columns
  JOURJR-65002NF
HPLC columns

Analytical, 2,10 mm, 50 mm

 
 
   
1 * 1 Pce
 

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
 
  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Length:
    50 mm
  • Seq:
    02