Order Entry
France
Orders LinkContactUsLinkComponent
HPLC columns

HPLC columns

Analytical, 2,10 mm, 30 mm
Catalog # JOURJR-65001NF
Supplier:   VICI JOUR RESEARCH
HPLC columns
Catalog # JOURJR-65001NF
Supplier:  VICI JOUR RESEARCH
HPLC columns

Analytical, 2,10 mm, 30 mm

Price
Inquire For Price
Log in for availability
1 * 1 Pce
Product Details & Documents

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
Specifications
  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Length:
    30 mm
  • Seq:
    01