Order Entry
France
Orders LinkContactUsLinkComponent
HPLC columns

HPLC columns

Analytical, 4,00 mm, 50 mm
Catalog # JOURJR-68181NF
Supplier:   VICI JOUR RESEARCH
HPLC columns
Catalog # JOURJR-68181NF
Supplier:  VICI JOUR RESEARCH
HPLC columns

Analytical, 4,00 mm, 50 mm

Price
234,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents

These PEEK columns are designed for naro-bore applications with small amount of samples.

  • Biocompatible
  • High pressure applications
  • Packing devices available
Specifications
  • Column Format:
    Analytical
  • Ø int.:
    4,00 mm
  • Length:
    50 mm
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.