(±)-2-Hexanol 99%
Glass bottle, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
- CAS Number:626-93-7
- Formula:C₆H₁₄O
- MDL Number:MFCD00004585
- Molecular Weight:102.18 g/mol
- Boiling Pt:139 °C (760 mmHg)
- Melting Pt:–23 °C
- Density:0.81 g/cm³
- Flash Pt:41 °C
- Storage Temperature:Ambient
- EINECS:210-971-4
- UN:2282
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4125 to 1.4155 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:10 g



