PISTON SEAL FOR IC 1 * 1 items
Catalog # METR6.2741.020
Supplier: METROHM
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Accessories for PIPETBOY acu / pro
Supplier: Integra