Order Entry
France
Orders LinkContactUsLinkComponent
First aid kit components

First aid kit components

Alcoholic spray, 70°, 50 ml
Catalog # LBES8170102
First aid kit components
Catalog # LBES8170102
First aid kit components
First aid kit components

Alcoholic spray, 70°, 50 ml

Price
24,30 €
Log in for availability
1 * 1 Pce
Product Details & Documents

Sprays, gels and creams for wound disinfection and burn treatment.

Specifications
  • Description:
    Alcoholic spray, 70°, 50 ml
  • Seq:
    03