Disposable cuvettes
Disposable cuvettes, 2,4 – 4,5 ml, 10 mm, Plastic
JENW83070-32
Plastic disposable cuvettes for liquid samples in spectrophotometry.
- Path length:10 mm
- Volume:2,4 – 4,5 ml
- Window material:Plastic
- Description:Disposable cuvettes
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



