BUFFER SOLUTION PH7 1 * 1 L
Catalog # FFCHFF1721000
Supplier: FF CHEMICALS
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Disposable cuvettes
Supplier: Brand
VWR® Low Nitrogen Content Filters, Grade 521
Supplier: VWR Collection
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
BD Microlance™ 3 Needles
Supplier: BD Medical
Acidic cleaner and neutralizer, borer 25 ORGANACID
Supplier: Borer Chemie
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Cuvettes and tubes for Picco, Multy, Nova and Prove
Supplier: Merck
Measuring beakers with handle
Supplier: VITLAB
Bromocresol green 0,04% in ethanol, TECHNICAL
Supplier: VWR Chemicals
BUFFER SOLUTION PH10 1 * 1 L
Supplier: FF CHEMICALS
Hydrochloric acid 0.1 mol/l (0.1 N)
Supplier: FF CHEMICALS
BUFFER SOLUTION PH4 1 * 1 L
Supplier: FF CHEMICALS
BUFFER SOLUTION PH2 1 * 1 L
Supplier: FF CHEMICALS
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Supplier: FF CHEMICALS
VWR®, Speciality pH Electrodes
Supplier: VWR Collection
BUFFER SOLUTION PH12 1 * 1 L
Supplier: FF CHEMICALS