BUFFER SOLUTION PH2 1 * 1 L
Catalog # FFCHFF1611000
Supplier: FF CHEMICALS
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Latex gloves, Semperguard®
Supplier: SEMPERIT TECHN PROD
pH buffers, ready to use
Supplier: WTW