Order Entry
France
Orders LinkContactUsLinkComponent
 

Antibiotic assay discs

Antibiotic assay discs, Ø 13 mm, 1.000
Catalog # PRATA013242
Supplier:   PRAT DUMAS
Antibiotic assay discs
Catalog # PRATA013242
Supplier:  PRAT DUMAS
CAS Number:  
Antibiotic assay discs
Antibiotic assay discs

Antibiotic assay discs, Ø 13 mm, 1.000

CAS Number:  
Price
89,70 €
Log in for availability
1 * 1.000 Pce
Product Details & Documents

For determining antibiotic suited to the treatment of an infection and for checking their sensitivity to antibiotics by means of the inhibition zone determination method.

  • Very thick cotton fibre papers, for the absorption of test solutions in antibiotic assays
  • 420 g/m² weight
Specifications

Specifications

  • Pk:
    1.000
  • Description:
    Antibiotic assay discs, Ø 13 mm

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.