Order Entry
France
Orders LinkContactUsLinkComponent
Antibiotic assay discs

Antibiotic assay discs

Antibiotic assay discs, Ø 12 mm, 1.000
Catalog # PRATA012242
Supplier:   PRAT DUMAS
Antibiotic assay discs
Catalog # PRATA012242
Supplier:  PRAT DUMAS
CAS Number:  
Antibiotic assay discs
Antibiotic assay discs

Antibiotic assay discs, Ø 12 mm, 1.000

CAS Number:  
Price
97,50 €
Log in for availability
1 * 1.000 Pce
Product Details & Documents

For determining antibiotic suited to the treatment of an infection and for checking their sensitivity to antibiotics by means of the inhibition zone determination method.

  • Very thick cotton fibre papers, for the absorption of test solutions in antibiotic assays
  • 420 g/m² weight

Documents

Specifications

Specifications

  • Pk:
    1.000
  • Description:
    Antibiotic assay discs, Ø 12 mm

Specifications