Jump to:
Product Details & Documents
Fast nano-LC for proteins and peptides. Chromolith® CapRod® capillaries combine the speed of monolithic silica technology with the sensitivity of nano-LC, thus enabling high speed high sensitivity LC-MS analytics.
- High flow rates at low pressure
- Long column lifetime
- Robust handling
Documents
Merck Chromolith® HPLC Columns
Product Literature
Specifications
- Column Format:Capillary
- Ø int.:0,10 mm
- Length:50 mm
- Seq:10
- Phase:RP-18e Trap



